Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 550 565 1/EA
Catalog No. 5000500099
Encompass_Preferred
ActinBrite 550/565 High Affinity Phalloidin Conjugate is a novel orange-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size
Catalog No. 50-005-00099 Supplier Biotium Supplier No. 00097T
ActinBrite 550/565 High Affinity Phalloidin Conjugate is a novel orange-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size