Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 550 565 1/EA
Supplier:  Biotium 00097
Encompass_Preferred
ActinBrite 550/565 High Affinity Phalloidin Conjugate is a novel orange-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size
Catalog No. 50-005-00150