Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 600 630 1/EA
Catalog No. 5000500129
Encompass_Preferred
ActinBrite 600/630 High Affinity Phalloidin Conjugate is a novel red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size
Catalog No. 50-005-00129 Supplier Biotium Supplier No. 00098T
ActinBrite 600/630 High Affinity Phalloidin Conjugate is a novel red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size