Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 610 630 1/EA
Catalog No. 5000501274
Encompass_Preferred
ActinBrite 610/630 High Affinity Phalloidin Conjugate is a novel red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size
Catalog No. 50-005-01274 Supplier Biotium Supplier No. 00098
ActinBrite 610/630 High Affinity Phalloidin Conjugate is a novel red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size