Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 645 665 1/EA
Catalog No. 5000500679
Encompass_Preferred
ActinBrite 645/665 High Affinity Phalloidin Conjugate is a novel far-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size
Catalog No. 50-005-00679 Supplier Biotium Supplier No. 00101T
ActinBrite 645/665 High Affinity Phalloidin Conjugate is a novel far-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 50 U trial size