Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 645 665 1/EA
Supplier:  Biotium 00101
Encompass_Preferred
ActinBrite 645/665 High Affinity Phalloidin Conjugate is a novel far-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size
Catalog No. 50-005-00131