Learn More
Biotium Toxins Receptor Probes Cell and Organelle Stains Cytoskeletal Stains ActinBrite 665 690 1/EA
Catalog No. 5000499440
Encompass_Preferred
ActinBrite 665/690 High Affinity Phalloidin Conjugate is a novel far-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size
Catalog No. 50-004-99440 Supplier Biotium Supplier No. 00099
ActinBrite 665/690 High Affinity Phalloidin Conjugate is a novel far-red fluorescent phalloidin designed to preserve binding affinity for F-actin resulting in bright staining that can be imaged more than one month after staining with minimal loss of signal or specificity 300 U size