Learn More
Biotium Toxins Receptor Probes Fluorescent Proteins Nucleotides Other Conjugates Cholera Toxin and Transferrin Conjugates CF740 1/EA
Catalog No. 5000500133
Encompass_Preferred
Cholera toxin B subunit (CTB) binds GM1 ganglioside Fluorescent CTB is used to label lipid rafts and as a neuronal retrograde tracer Biotiums CF dyes have superior brightness and photostability compared to other dyes Near-infrared CF740 dye has Ex/Em at 742/767 nm 100 ug
Catalog No. 50-005-00133 Supplier Biotium Supplier No. 29127
Cholera toxin B subunit (CTB) binds GM1 ganglioside Fluorescent CTB is used to label lipid rafts and as a neuronal retrograde tracer Biotiums CF dyes have superior brightness and photostability compared to other dyes Near-infrared CF740 dye has Ex/Em at 742/767 nm 100 ug