Learn More
Biotium Toxins Receptor Probes Fluorescent Proteins Nucleotides Other Conjugates Cholera Toxin and Transferrin Conjugates Unlabeled 1/EA
Supplier: Biotium 29130100UG
This product is recombinant Cholera Toxin Subunit B expressed from E coli cells It is supplied lyophilized and binds GM1 ganglioside This product is dye conjugation compatible and fluorescent CTB can be used to label lipid rafts and as a neuronal retrograde tracer to cells Unit size 100 ug