Learn More
Biotium Toxins Receptor Probes Fluorescent Proteins Nucleotides Other Conjugates Cholera Toxin and Transferrin Conjugates Unlabeled 1/EA
Supplier:  Biotium 291302MG
Encompass_Preferred
This product is recombinant Cholera Toxin Subunit B expressed from E coli cells It is supplied lyophilized and binds GM1 ganglioside This product is dye conjugation compatible and fluorescent CTB can be used to label lipid rafts and as a neuronal retrograde tracer to cells Unit size 2 mg
Catalog No. 50-005-00135