Learn More
Genscript Corporation ToxinSensor Chromogenic LAL E
Supplier:  Genscript Corporation L00350C
Encompass
Genscript ToxinSensor Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs biological products and medical devices The system is not intended for detection of endotoxin in a licensed reagent clinical samples or the diagnosis of human disease A measurable endotoxin concentration range from 0 01 to 1 EU/ml can be achieved Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus)
Catalog No. 50-000-59540