Learn More
Genscript Corporation ToxinSensor Endotoxin-free Tu
Supplier:  Genscript Corporation M010725
Encompass
GenScript supplies 2 5 ml endotoxin-free tubes (internal diameter 8 mm external diameter 10 mm height 50 mm) with plain end made of research-grade glass GenScript s Endotoxin-free tubes can be applied to dilute or aliquot sample with an especial requirement of endotoxin level ( 0 005 EU/ml) br GenScript Endotoxin-free tubes are provided in encapsulation with pyrogen-free aluminum foil (folded into a package) which sufficiently inhibits the introduction of any contamination such as RNase/DNase br/
Catalog No. 50-000-58077