Learn More
Genscript Corporation ToxinSensor Single Tests Kit
Supplier:  Genscript Corporation L0085920
Encompass
GenScript ToxinSensor Single Tests Kit with Standard is designed to be a qualitative In Vitro end-point endotoxin test for human and animal parenteral drugs biological products and medical devices Tachypleus Amebocyte Lysate (TAL) as supplied is to be reconstituted with sample or control directly After incubation for 1 hour and in the presence of endotoxin gelation occurs in the absence of endotoxin gelation does not occur GenScript supply a series of ToxinSensorTM Single Tests Kits with Standard with different sensitivity (0 03 EU/ml 0 06 EU/ml 0 125 EU/ml and 0 25 EU/ml)
Catalog No. 50-000-57418