Learn More
TARGETMOL CHEMICALS INC TOXOFLAVIN 10MG
SDP
Catalog No. 502581122
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Toxoflavin (Xanthothricin) is an antagonist of transcription factor 4 (TCF4)/(beta)-catenin complex. Toxoflavin also acts as an inhibitor of KDM4A. purity: 99%
Catalog No. 50-258-1122 Supplier TARGETMOL CHEMICALS INC Supplier No. T1714210MG
Also available in 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Toxoflavin (Xanthothricin) is an antagonist of transcription factor 4 (TCF4)/(beta)-catenin complex. Toxoflavin also acts as an inhibitor of KDM4A. purity: 99%