Learn More
Bioss Toxoplasma gondii
SDP
Catalog No. 5000066129
Encompass_Preferred
Toxoplasma gondii Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66129 Supplier Bioss Supplier No. BSM72437M
Toxoplasma gondii Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide