Learn More
Alpco Ltd Toxoplasma Gondii IgA ELISA 96 Wells 20 L
SDP
Supplier:  Alpco Ltd 20TGAHUE01
Encompass_Preferred
The Toxoplasma gondii IgA Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgA-class antibodies to Toxoplasma gondii in serum or plasma. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-130