Learn More
Drg International Inc DRG Toxoplasma Gondii IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3520R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgM-class antibodies to Toxoplasma gondii in human serum and plasma. This product is for Research Use Only. The DRG Toxoplasma gondii IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent to eliminate competitive inhibition from specific IgG. This pretreatment avoids false results. Microtiter wells, as a solid phase are coated with inactivated Toxoplasma gondii soluble antigen (strain RH). 
Catalog No. 50-194-7591