Learn More
ATCC Toxoplasma gondii; ME49
Catalog No. 502382743
Encompass
Toxoplasma gondii strain me49 is in haplogroup 2. This parasitic protozoan is propagated in Hs27 cells (ATCC CRL-1634) and can be used in food and waterborne pathogen research and opportunistic pathogen research.
Catalog No. 50-238-2743 Supplier ATCC Supplier No. 50611
Toxoplasma gondii strain me49 is in haplogroup 2. This parasitic protozoan is propagated in Hs27 cells (ATCC CRL-1634) and can be used in food and waterborne pathogen research and opportunistic pathogen research.