Learn More
Bioworld Toxoplasma Gondii P24 (GRA 1) (Recombinant), 100 μg
Supplier:  Bioworld 220607261
Encompass_Preferred
Recombinant Toxoplasma Gondii Dense granule protein 1 (GRA1/p24). Source: E.coli Description : The protein contains the P24 (GRA1) immunodominant regions, amino acids:(57-149). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining). Protein conc: 1mg/ml in 50 mM Tris-HCl, 1.5 M urea and 50% glycerol. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of Toxoplasma gondii-infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for detection of Toxoplasma gondii- with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-824