Learn More
Bioworld Toxoplasma Gondii P29 (GRA 7) (Recombinant), 100 μg
Supplier:  Bioworld 220607271
Encompass_Preferred
Recombinant Toxoplasma Gondii Granule Protein 7 (GRA7/p29) Source: E.coli Description : The protein contains the P29 (GRA7) immunodominant regions, amino acids:(24-100). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining). Protein conc: 1mg/ml in 50 mM Tris-HCl, 1.5 M urea and 50% glycerol. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of Toxoplasma gondii-infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for detection of Toxoplasma gondii- with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-825