Learn More
Cell Signaling Technology Tox and Tox2 Matched Antibody Pair, Rabbit Monoclonal, Human, Mouse, Rat Reactivity, 1 Kit
SDP
Catalog No. 503134303
Encompass
The Tox/Tox2 Matched Antibody Pair consists of capture and detection antibodies designed for use in immunoassay technologies, such as high-throughput ELISA platforms. This kit is ideal for applications requiring custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg each of rabbit monoclonal capture and detection antibodies
  • BSA and azide free formulation
  • Validated for reactivity in human, mouse, and rat models
  • Concentration of 1 mg/mL
  • Designed for use with various immunoassay platforms including AlphaScreen, AlphaLISA, LANCE, HTRF, and Luminex
Catalog No. 50-313-4303 Supplier Cell Signaling Technology Supplier No. 76385S
The Tox/Tox2 Matched Antibody Pair consists of capture and detection antibodies designed for use in immunoassay technologies, such as high-throughput ELISA platforms. This kit is ideal for applications requiring custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg each of rabbit monoclonal capture and detection antibodies
  • BSA and azide free formulation
  • Validated for reactivity in human, mouse, and rat models
  • Concentration of 1 mg/mL
  • Designed for use with various immunoassay platforms including AlphaScreen, AlphaLISA, LANCE, HTRF, and Luminex