Learn More
Medchemexpress LLC HY-103248 5mg Medchemexpress, Toyocamycin CAS:606-58-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1032485MG
Encompass_Preferred
Medchemexpress, HY-103248 5mg Toyocamycin CAS:606-58-6 Toyocamycin (Vengicide) is an adenosine analog produced by Actinomycete, acts as an XBP1 inhibitor, inhibits IRE1α-induced ATP-dependent XBP1 mRNA cleavage, with an IC 50 of 80 nM. Toyocamycin (Vengicide) induces apoptosis. Toyocamycin (Vengicide) shows no effect on IRE1α auto-phosphorylation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2240