Learn More
Cayman Chemical ToyocamycInhydrate 10mg
SDP
Catalog No. 502968422
Encompass_Preferred
A natural adenosine analog that prevents IRE1α-induced mRNA cleavage (IC50 = 80 nM) and inhibits constitutive activation of XBP1 in multiple myeloma cell lines; used to study IRE1α action in the endoplasmic reticulum stress response
Catalog No. 50-296-8422 Supplier Cayman Chemical Supplier No. 1737110 MG
A natural adenosine analog that prevents IRE1α-induced mRNA cleavage (IC50 = 80 nM) and inhibits constitutive activation of XBP1 in multiple myeloma cell lines; used to study IRE1α action in the endoplasmic reticulum stress response