Learn More
Tosoh Bioscience LLC TOYOPEARL AF-CHELATE-650M 25ML
Supplier:  Tosoh Bioscience LLC 0014475
Encompass
Media, Ready-to-Use Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Chelate-650M, 65 micrometer; Matrix: Polymer; Container Size: 25mL
Catalog No. 50-851-367