Learn More
Tosoh Bioscience LLC TOYOPEARL AF-EPOXY-650M 100G
Supplier:  Tosoh Bioscience LLC 0008038
Encompass
Media, Activated Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Epoxy-650M, 65 micrometer; Matrix: Polymer; 100g
Catalog No. 50-851-257