Learn More
Tosoh Bioscience LLC TOYOPEARL AF-EPOXY-650M 10G
Supplier: Tosoh Bioscience LLC 0008000
Media, Activated Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Epoxy-650M, 65 micrometer; Matrix: Polymer; 10g