Learn More
Tosoh Bioscience LLC TOYOPEARL AF-FORMYL-650M 10ML
Supplier:  Tosoh Bioscience LLC 0043413
Encompass
Media, Reactive Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Formyl-650M, 65 micrometer; Matrix: Polymer; Container Size: 10mL
Catalog No. 50-851-752