Learn More
Tosoh Bioscience LLC TOYOPEARL AF-FORMYL-650M 25ML
Supplier:  Tosoh Bioscience LLC 0008004
Encompass
Media, Reactive Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Formyl-650M, 65 micrometer; Matrix: Polymer; Container Size: 25mL
Catalog No. 50-851-239