Learn More
Tosoh Bioscience LLC TOYOPEARL AF-TRESYL-650M 200G
Supplier:  Tosoh Bioscience LLC 0014905
Encompass
Media, Activated Bulk Resins; For Affinity Chromatography; Tosoh Toyopearl AF-Tresyl-650M, 65 micrometer; Matrix: Polymer; 200g
Catalog No. 50-851-378
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.