Learn More
Tosoh Bioscience LLC TOYOPEARL AFFIPAK 3X10ML
Supplier:  Tosoh Bioscience LLC 0043410
Encompass
Media; Process Resins; LabPak Sampler for Affinity Chromatography; Tosoh Toyopearl AFFIPAK (AF-Amino, Carboxy, Formyl-650M), 65 micrometer; Matrix: Polymer; 3 x 10mL
Catalog No. 50-851-749