Learn More
Tosoh Bioscience LLC TOYOPEARL BUTYL-600M 100ML
Supplier:  Tosoh Bioscience LLC 0021449
Encompass
Media, Process Resins; For Hydrophobic Interaction Chromatography; Tosoh Toyopearl Butyl-600M, 65 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-677