Learn More
Tosoh Bioscience LLC TOYOPEARL BUTYL-650S 100ML
Supplier:  Tosoh Bioscience LLC 0007476
Encompass
Media, Bulk Resins; For Hydrophobic Interaction Chromatography; Tosoh Toyopearl Butyl-650S, 35 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-206