Learn More
Tosoh Bioscience LLC TOYOPEARL CM-650M 250ML 65UM
Supplier:  Tosoh Bioscience LLC 0007475
Encompass
Media, Bulk Cation Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl CM-650M, 65 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-205