Learn More
Tosoh Bioscience LLC TOYOPEARL CM-650S 25ML
Supplier:  Tosoh Bioscience LLC 0019803
Encompass_Preferred
Media, Bulk Cation Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl CM-650S, 35 micrometer; Matrix: Polymer; Container Size: 25mL
Catalog No. 50-851-589