Learn More
Tosoh Bioscience LLC TOYOPEARL DEAE-650C 250ML
Supplier:  Tosoh Bioscience LLC 0007988
Encompass
Media, Process Resins; Bulk Anion Exchangers; For Ion Exchange Chromatography; Tosoh Toyopearl DEAE-650C, 100 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-233