Learn More
Tosoh Bioscience LLC TOYOPEARL DEAE-650M 100ML
Supplier:  Tosoh Bioscience LLC 0043201
Encompass_Preferred
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl DEAE-650M, 65 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-727