Learn More
Tosoh Bioscience LLC TOYOPEARL GIGACAP Q-650S 25ML
Catalog No. 50486531 Shop All Tosoh Bioscience LLC Products
Encompass
LC Ion Exchange Media, Toyopearl GigaCap Q-650S, 25 mL, 35 um
Catalog No. 50-486-531 Supplier Tosoh Bioscience LLC Supplier No. 0022881
LC Ion Exchange Media, Toyopearl GigaCap Q-650S, 25 mL, 35 um