Learn More
Tosoh Bioscience LLC TOYOPEARL HICPAK-C 3X25ML
Supplier:  Tosoh Bioscience LLC 0043125
Encompass
Media, Process Resins; LabPak Sampler for Hydrophobic Interaction Chromatography; Tosoh Toyopearl HICPAK-C (Phenyl, Butyl, Hexyl-650C), 100 micrometer; Matrix: Polymer; 3 x 25mL
Catalog No. 50-851-717