Learn More
Tosoh Bioscience LLC TOYOPEARL HICPAK HP 3X25ML
Supplier:  Tosoh Bioscience LLC 0043150
Encompass
Media, Process Resins; LabPak Sampler for Hydrophobic Interaction Chromatography; Tosoh Toyopearl HICPAK HP (Ether, Phenyl, Butyl-650S) , 35 micrometer; Matrix: Polymer; 3 x 25mL
Catalog No. 50-851-720