Learn More
Tosoh Bioscience LLC TOYOPEARL HW-40C 500ML 75UM
Supplier:  Tosoh Bioscience LLC 0007449
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-40C, 75 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-186