Learn More
Tosoh Bioscience LLC TOYOPEARL HW-40EC 500ML
Supplier:  Tosoh Bioscience LLC 0007450
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-40EC, 200 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-187