Learn More
Tosoh Bioscience LLC TOYOPEARL HW-50F 150ML 45UM
Supplier: Tosoh Bioscience LLC 0019810
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-50F, 45 micrometer; Matrix: Polymer; Container Size: 150mL