Learn More
Tosoh Bioscience LLC TOYOPEARL HW-50F 500ML 45UM
Supplier:  Tosoh Bioscience LLC 0007453
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-50F, 45 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-190