Learn More
Tosoh Bioscience LLC TOYOPEARL HW-50S 500ML 30UM
Supplier:  Tosoh Bioscience LLC 0007452
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-50S, 30 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-189