Learn More
Tosoh Bioscience LLC TOYOPEARL HW-55F 500ML 45UM
Supplier:  Tosoh Bioscience LLC 0007457
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-55F, 45 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-193