Learn More
Tosoh Bioscience LLC TOYOPEARL HW-55S 150ML 30UM
Supplier:  Tosoh Bioscience LLC 0019813
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-55S, 30 micrometer; Matrix: Polymer; Container Size: 150mL
Catalog No. 50-851-599