Learn More
Tosoh Bioscience LLC TOYOPEARL HW-65C 150ML 75UM
Supplier:  Tosoh Bioscience LLC 0021481
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-65C, 75 micrometer; Maxtrix: Polymer; Container Size: 150mL
Catalog No. 50-851-688