Learn More
Tosoh Bioscience LLC TOYOPEARL HW-65C 75UM 500ML
Supplier:  Tosoh Bioscience LLC 0007466
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-65C, 75 micrometer; Matrix: Polymer; Container Size: 500mL
Catalog No. 50-851-197