Learn More
Tosoh Bioscience LLC TOYOPEARL HW-65F 150ML 45UM
Supplier:  Tosoh Bioscience LLC 0019814
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-65F, 45 micrometer; Matrix: Polymer; Container Size: 150mL
Catalog No. 50-851-600