Learn More
Tosoh Bioscience LLC TOYOPEARL HW-65S 500ML 30UM
Supplier: Tosoh Bioscience LLC 0007464
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-65S, 30 micrometer; Matrix: Polymer; Container Size: 500mL